Diphenyl phosphate 99%

Leverandør: Thermo Fisher Scientific

Synonymer: Diphenyl hydrogen phosphate

225690050 225690250
ACRO225690050EA 910 NOK
ACRO225690050 ACRO225690250
Diphenyl phosphate 99%
Diphenyl phosphate
Formel: C₁₂H₁₁O₄P
MW: 250.19 g/mol
Smeltepunkt: 63…70 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003033
CAS-nummer: 838-85-7
EINECS: 212-657-2

Bestill nå

Spesifikasjon Testresultater

Appearance White Powder or crystals
Infrared spectrum Conforms
Melting point 62 to 70 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR