Lactulose 99+%

Leverandør: Thermo Fisher Scientific

225930250 225930050 225931000
ACRO225930250EA 2300 NOK
ACRO225930250 ACRO225930050 ACRO225931000
Lactulose 99+%
Lactulose
Formel: C₁₂H₂₂O₁₁
Smeltepunkt: 173…178 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063276
CAS-nummer: 4618-18-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 % (1 g, 105°C, 3 h)
HPLC ≥99.0 % (ELSD)
Specific optical rotation -46° to -50° (20°C, 589 nm) (c=1, H2O, Ammonia)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR