2'-Deoxycytidine 99+%

Leverandør: Thermo Fisher Scientific

226720010 226722500 226725000 226720050 226720250
ACRO226720010EA 1200 NOK
ACRO226720010 ACRO226722500 ACRO226725000 ACRO226720050 ACRO226720250
2'-Deoxycytidine 99+%
2'-Deoxycytidine
Formel: C₉H₁₃N₃O₄
Smeltepunkt: 209…211 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006547
CAS-nummer: 951-77-9
EINECS: 213-454-1

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤8 %
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR