D-(-)-Valine 98+%

Leverandør: Thermo Fisher Scientific

227220250 227220050
ACRO227220250EA 1775 NOK
ACRO227220250 ACRO227220050
D-(-)-Valine 98+%
D-(-)-Valine
Formel: C₅H₁₁NO₂
Kokepunkt: 217 °C (1013 hPa)
Smeltepunkt: 302…303 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064219
CAS-nummer: 640-68-6
EINECS: 211-368-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Loss on drying ≤0.5 % (105°C, 4 hrs)
Sulfated ash ≤0.2 %
TLC ≤1 % L-enantiomer
Specific optical rotation -26° to -29° (20°C, 589 nm) (c=5, 5 N HCl)
Titration with HClO4 98.0 to 102.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR