Puromycin dihydrochloride hydrate 99%

Leverandør: Thermo Fisher Scientific
Warning

227420100 227420500 227422500
ACRO227420100EA 1100 NOK
ACRO227420100 ACRO227420500 ACRO227422500
Puromycin dihydrochloride hydrate 99%
Puromycin dihydrochloride hydrate
Formel: C₂₂H₂₉N₇O₅·2HCl·nH₂O
MW: 544.44 g/mol (anhy)
Smeltepunkt: 168…178 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00150080
CAS-nummer: 58-58-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to light beige Powder
Infrared spectrum Conforms
Water ≤12 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR