ACES 99%, extra pure

Leverandør: Thermo Fisher Scientific

Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

228000250 228001000 228002500
ACRO228000250EA 1150 NOK
ACRO228000250 ACRO228001000 ACRO228002500
ACES 99%, extra pure
ACES
Formel: H₂NCOCH₂NHCH₂CH₂SO₃H
Smeltepunkt: 270…295 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008030
CAS-nummer: 7365-82-4
EINECS: 230-908-4
Merck Indeks: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV at 280 nm A: ≤0.02
pH 3.6 to 4.4 (1 % solution)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR