Cytochalasin B

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione

228090010 228090050 228090250
ACRO228090010EA 1225 NOK
ACRO228090010 ACRO228090050 ACRO228090250
Cytochalasin B
Cytochalasin B
Formel: C₂₉H₃₇NO₅
MW: 479.62 g/mol
Smeltepunkt: 218…223 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077704
CAS-nummer: 14930-96-2
EINECS: 239-000-2
FN: 1544
ADR: 6.1 II

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
HPLC ≥98 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR