Digoxin 95%

Leverandør: Thermo Fisher Scientific
Danger

228120010 228120050
ACRO228120010EA 1650 NOK
ACRO228120010 ACRO228120050
Digoxin 95%
Digoxin
Formel: C₄₁H₆₄O₁₄
Smeltepunkt: 248…250 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003674
CAS-nummer: 20830-75-5
EINECS: 244-068-1
FN: 3462
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 % (500 mg) (vacuum)
Sulfated ash ≤0.1 %
HPLC ≥94 % (on dried substance)
Specific optical rotation +10° to +13° (20°C, 589 nm) (c=2, anhydr. pyridine) (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR