Lys Grønn SF Gulaktig, pure, high purity biological stain

Leverandør: Thermo Fisher Scientific

229770100 229771000
ACRO229770100EA 1375 NOK
ACRO229770100 ACRO229771000
Lys Grønn SF Gulaktig, pure, high purity biological stain
Lys Grønn SF Gulaktig
Formel: C₃₇H₃₄N₂Na₂O₉S₃
Smeltepunkt: 288 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012121
CAS-nummer: 5141-20-8
EINECS: 225-906-5
Merck Indeks: 13,05506

Bestill nå

Spesifikasjon Testresultater

Appearance Purple Powder or crystals
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤10 % (1 g, 110°C, 2 h)
UV Lambda max 629 to 634 nm (H2O, 5 mg/l, 1 cm)
Absorption Ratio P-15/P+15 0.96 to 1.16
Titration ≥60 % (As is)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR