Sodium taurocholate hydrate 98%

Leverandør: Thermo Fisher Scientific
Warning

230370010 230370050 230370250 230371000
ACRO230370010EA 860 NOK
ACRO230370010 ACRO230370050 ACRO230370250 ACRO230371000
Sodium taurocholate hydrate 98%
Sodium taurocholate hydrate
Formel: C₂₆H₄₆NNaO₈S
Smeltepunkt: 230 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150819
CAS-nummer: 345909-26-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to cream Powder
Infrared spectrum Conforms
Water ≤7 %
TLC ≥97.5 %
Residue after ignition 11.0 % to 14.0 %
Specific optical rotation +20.0° to +25.8° (20°C, 589 nm) (c=3, water)
Heavy metals (as Pb) ≤20 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR