Ethyl cellulose, 300 cps 48% (ethoxyl content)

Leverandør: Thermo Fisher Scientific

232705000 232700051
ACRO232705000EA 2525 NOK
ACRO232705000 ACRO232700051
Ethyl cellulose, 300 cps 48% (ethoxyl content)
Ethyl cellulose

CAS No.: 9004-57-3

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00131037
CAS-nummer: 9004-57-3

Bestill nå

Spesifikasjon Testresultater

Appearance White Granular powder
Infrared spectrum Conforms
Assay 48 to 49.5 % (ethoxyl)
Viscosity 250 to 350 cP (5 % in 80/20 toluene/ethanol at 25°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR