O-Phenetidine 99%

Leverandør: Thermo Fisher Scientific
Danger

233480050 233485000
ACRO233480050EA 355 NOK
ACRO233480050 ACRO233485000
O-Phenetidine 99%
O-Phenetidine
Formel: C₈H₁₁NO
Kokepunkt: 229 °C (1013 hPa)
Smeltepunkt: –20 °C
Tetthet: 1,05 g/cm³
Flash punkt: 80 °C
MDL nummer: MFCD00007689
CAS-nummer: 94-70-2
EINECS: 202-356-4
FN: 2311
ADR: 6.1,III
Merck Indeks: 12,07372

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to red Liquid
Infrared spectrum Conforms
Refractive index 1.5540 to 1.5560 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR