5-Nitroantranilinsyre 98%

Leverandør: Thermo Fisher Scientific
Warning

245950050 245950250
ACRO245950050EA 510 NOK
ACRO245950050 ACRO245950250
5-Nitroantranilinsyre 98%
5-Nitroantranilinsyre
Formel: C₇H₆N₂O₄
Smeltepunkt: 278 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017039
CAS-nummer: 616-79-5
EINECS: 210-493-6

Bestill nå

Spesifikasjon Testresultater

Appearance Bright yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR