Promethazine hydrochloride

Leverandør: Thermo Fisher Scientific
Warning

254580050 254580250 254581000
ACRO254580050EA 625 NOK
ACRO254580050 ACRO254580250 ACRO254581000
Promethazine hydrochloride
Promethazine hydrochloride
Formel: C₁₇H₂₀N₂S
Smeltepunkt: 227…230 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012652
CAS-nummer: 58-33-3
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 4 hr)
Solubility (10% in water) Clear colorless to light yellow
Titration with HClO4 97.5 to 101.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR