alpha-Apo-oxytetracycline, can be used as secondary standard

Leverandør: Thermo Fisher Scientific

257721000
ACRO257721000EA 2675 NOK
ACRO257721000
alpha-Apo-oxytetracycline, can be used as secondary standard
Standarder Organiske standarder

CAS No.: 18695-01-7


MDL No.: MFCD00062827
GHS Symbols: GHS07, GHS08,
Melting Point: 200 °C (D)
Appearance: Khaki Crystalline powder
Storage Temperature: Ambient
Signal word: Warning

Bestill nå

SPESIFIKASJONER

Beskrivelse alpha-Apo-oxytetracycline
Environmentally Preferable
CAS-nr 18695-01-7
Formel C₂₂H₂₂N₂O₈

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR