beta-Apo-oxytetracycline, can be used as secondary standard

Leverandør: Thermo Fisher Scientific

257731000
ACRO257731000EA 2875 NOK
ACRO257731000
beta-Apo-oxytetracycline, can be used as secondary standard
Standarder Organiske standarder

CAS No.: 18751-99-0


MDL No.: MFCD00062827
GHS Symbols: GHS08
Melting Point: 200 to 204 °C (D)
Appearance: Yellow-brown to brown Powder
Storage Temperature: Ambient
Signal word: Warning

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR