L-(-)-α-Amino-γ-butyrolactone hydrobromide 99%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: L-(-)-α-Amino-γ-butyrolactone hydrobromide, (S)-2-amino-4-butyrolactone hydrobromide, (S)-(-)-α-Amino-γ-butyrolactone hydrobromide, (S)-3-Aminodihydrofuran-2(3H)-one hydrobromide

259940010 259940050
ACRO259940010EA 590 NOK
ACRO259940010 ACRO259940050
L-(-)-α-Amino-γ-butyrolactone hydrobromide 99%
L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide
Formel: C₄H₇NO₂·HBr
MW: 182.017 g/mol
Smeltepunkt: 236…238.9 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00674493
CAS-nummer: 15295-77-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Specific optical rotation -20.5° to -22.5° (20°C, 589 nm) (c=4, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR