Tris phosphate 98%

Leverandør: Thermo Fisher Scientific

264952500
ACRO264952500EA 4500 NOK
ACRO264952500
Tris phosphate 98%
Tris phosphate

Appearance: White Crystalline powder

Formel: C₄H₁₄NO₇P
Smeltepunkt: 168…172 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00043285
CAS-nummer: 6992-39-8

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 172 °C
Titration with NaOH ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR