2-Mercapto-5-methoxybenzimidazole 99+%

Leverandør: Thermo Fisher Scientific
Warning

275370250 275370050
ACRO275370250EA 1650 NOK
ACRO275370250 ACRO275370050
2-Mercapto-5-methoxybenzimidazole 99+%
2-Mercapto-5-methoxybenzimidazole
Formel: C₈H₈N₂OS
Kokepunkt: 400 °C (1013 hPa)
Smeltepunkt: 255…262 °C
MDL nummer: MFCD00134581
CAS-nummer: 37052-78-1
EINECS: 253-326-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 255 to 260 °C
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR