2-Aminobenzaldehyde 97%

Leverandør: Thermo Scientific
Warning

279660010
ACRO279660010EA 3275 NOK
ACRO279660010
2-Aminobenzaldehyde 97%
2-Aminobenzaldehyde
Formel: C₇H₇NO
MW: 121,14 g/mol
Kokepunkt: 80…85 °C (2 mmHg)
Smeltepunkt: 37 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00007709
CAS-nummer: 529-23-7

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 34 to 40 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR