Biphenylene 99+%

Leverandør: Thermo Fisher Scientific

279742500
ACRO279742500EA 2075 NOK
ACRO279742500
Biphenylene 99+%
Biphenylene
Formel: C₁₂H₈
Kokepunkt: 262 °C (1013 hPa)
Smeltepunkt: 109…112 °C
Tetthet: 1,23 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001110
CAS-nummer: 259-79-0
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow Needles
Infrared spectrum Conforms
Melting point 109 to 112 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR