Giemsa's stain solution biological stain

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

295591000 295595000
ACRO295591000EA 380 NOK
ACRO295591000 ACRO295595000
Giemsa's stain solution biological stain
Giemsa's stain
Tetthet: 0,99 g/cm³
Flash punkt: 15 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081642
CAS-nummer: 51811-82-6
EINECS: 257-438-2
FN: 1230
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Dark blue to dark purple Solution
Additional info Giemsa stain powder in MeOH, Glycerol mixture
UV Lambda max 646 to 651 nm (c=0.1, MeOH) Methylene blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR