2-Nitrophenyl isocyanate 99%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 1-Isocyanato-2-nitrobenzene, 2-Nitrophenylisocyanate

296690050 296690250 AC29669.1 AC29669EA AC29669.50
ACRO296690050EA 965 NOK
ACRO296690050 ACRO296690250 MAYBAC29669.1 MAYBAC29669EA MAYBAC29669.50
2-Nitrophenyl isocyanate 99%
2-Nitrophenyl isocyanate
Formel: C₇H₄N₂O₃
MW: 164.12 g/mol
Kokepunkt: 135…137 °C (17 mmHg)
Smeltepunkt: 38…41 °C
Flash punkt: >110 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007092
CAS-nummer: 3320-86-3
EINECS: 222-024-2
FN: 3335
ADR: 9 III

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 38 to 41 °C
GC ≥98.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR