4-Nitrophenyl isocyanate 98%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: 1-Isocyanato-4-nitrobenzene

296700050 296700250 AC29670.1 AC29670.50
ACRO296700050EA 625 NOK
ACRO296700050 ACRO296700250 MAYBAC29670.1 MAYBAC29670.50
4-Nitrophenyl isocyanate 98%
4-Nitrophenyl isocyanate
Formel: C₇H₄N₂O₃
MW: 164.12 g/mol
Kokepunkt: 277
Smeltepunkt: 56…62 °C
Flash punkt: >110 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007306
CAS-nummer: 100-28-7
EINECS: 202-836-3
FN: 3335
ADR: 9 III

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 56 to 62 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR