cis,cis-Muconic acid 98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: (2Z,4Z)-Hexa-2,4-dienedioic acid

297760025 297760100
ACRO297760025EA 1375 NOK
ACRO297760025 ACRO297760100
cis,cis-Muconic acid 98%
cis,cis-Muconic acid

CAS No.: 1119-72-8

Formel: C₆H₆O₄
Smeltepunkt: 194…195 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085266
CAS-nummer: 1119-72-8

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR