1,2-DiBrom-1-fenyletan 97%

Leverandør: Thermo Scientific
Danger

Synonymer: (1,2-Dibromoethyl)benzene

299305000
ACRO299305000EA 2575 NOK
ACRO299305000
1,2-DiBrom-1-fenyletan 97%
1,2-DiBrom-1-fenyletan
Formel: C₆H₅CHBrCH₂Br
MW: 263,96 g/mol
Kokepunkt: 139…141 °C (15 mmHg)
Smeltepunkt: 68…73 °C
Tetthet: 1,74 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000138
CAS-nummer: 93-52-7
EINECS: 202-253-4
FN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee6d6874d48c69b9de8b135a68f7f21d6b917e29&vid=a8a0e5da50d9cc34aa33f083227810709ad05908&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to cream to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 75 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR