Ethyl isocyanoacetate 97%

Leverandør: Thermo Fisher Scientific
Warning

299550050 299550250 299551000
ACRO299550050EA 1825 NOK
ACRO299550050 ACRO299550250 ACRO299551000
Ethyl isocyanoacetate 97%
Ethyl isocyanoacetate
Formel: C₅H₇NO₂
Kokepunkt: 194…196 °C (760 mmHg)
Tetthet: 1,03 g/cm³
Flash punkt: 84 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00000007
CAS-nummer: 2999-46-4
EINECS: 221-077-9
FN: 2810
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear amber to dark brown Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4250 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR