Ethephon(2-chloroethylphosphonic acid) 90% tech.

Leverandør: Thermo Fisher Scientific
Danger

301630050
ACRO301630050EA 11325 NOK
ACRO301630050
Ethephon(2-chloroethylphosphonic acid) 90% tech.
Ethephon(2-chloroethylphosphonic acid)
Formel: C₂H₆ClO₃P
Smeltepunkt: 74…75 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002142
CAS-nummer: 16672-87-0
FN: 2928
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 65 to 75 °C
Titration with NaOH ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR