4,4'-Diacetylbiphenyl 98%

Leverandør: Thermo Fisher Scientific
Warning

305440010 305440050
ACRO305440010EA 395 NOK
ACRO305440010 ACRO305440050
4,4'-Diacetylbiphenyl 98%
4,4'-Diacetylbiphenyl
Formel: C₁₆H₁₄O₂
Smeltepunkt: 192…196 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017248
CAS-nummer: 787-69-9
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to tan Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 197 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR