Ethyl-2-bromobenzoate 98%

Leverandør: Thermo Scientific
Warning

Synonymer: Ethyl 2-bromobenzoate, 2-Bromobenzoic acid ethyl ester

308530250
ACRO308530250EA 990 NOK
ACRO308530250
Ethyl-2-bromobenzoate 98%
Ethyl-2-bromobenzoate
Formel: C₉H₉BrO₂
MW: 229,07 g/mol
Kokepunkt: 117 °C (17 mmHg)
Tetthet: 1,44 g/cm³
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00015443
CAS-nummer: 6091-64-1
EINECS: 228-034-3

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5450 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR