L-Aspartyl-L-phenylalanine

Leverandør: Thermo Fisher Scientific

310021000 310025000
ACRO310021000EA 1375 NOK
ACRO310021000 ACRO310025000
L-Aspartyl-L-phenylalanine
Proteiner

CAS No.: 13433-09-5


  • Infrared spectrum: Conforms
  • Appearance of solution: (5% in CH₃COOH 80%) clear colorless
  • Specific optical rotation: 10.0 to 13.0° (24 °C, 589 nm) (c=1, 0.5 N HCl)
  • Melting point: 236 to 239 °C

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR