Cobalt(II) fluoride 99%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Cobalt difluoride, Cobaltous fluoride

311270250 311270050
ACRO311270250EA 1400 NOK
ACRO311270250 ACRO311270050
Cobalt(II) fluoride 99%
Cobalt(II) fluoride
Formel: CoF₂
MW: 96.93 g/mol
Kokepunkt: 1400 °C (760 mmHg)
Smeltepunkt: 1200 °C
Tetthet: 4.46
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010941
CAS-nummer: 10026-17-2
EINECS: 233-061-9
FN: 2923
ADR: 8 II
Merck Indeks: 13,02465

Bestill nå

Spesifikasjon Testresultater

Appearance Pink Powder
Assay ≥98.5 % (ex F)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR