orto-Borsyre 99.99% (trace metals basis), extra pure

Leverandør: Thermo Fisher Scientific
Danger

315181000 315180025 315185000
ACRO315181000EA 1200 NOK
ACRO315181000 ACRO315180025 ACRO315185000
orto-Borsyre 99.99% (trace metals basis), extra pure
orto-Borsyre
Formel: B(OH)₃
Kokepunkt: 185 °C (1013 hPa)
Smeltepunkt: 169 °C (dec.)
Tetthet: 1,44 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011337
CAS-nummer: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Indeks: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤100 ppm
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR