Diphenylphosphine 95%

Leverandør: Thermo Fisher Scientific
Danger

320970050 320970250
ACRO320970050EA 1200 NOK
ACRO320970050 ACRO320970250
Diphenylphosphine 95%
Diphenylphosphine
Formel: C₁₂H₁₁P
Flash punkt: 110 °C
MDL nummer: MFCD00003040
CAS-nummer: 829-85-6
EINECS: 212-591-4
FN: 2845
ADR: 4.2,I

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6250 to 1.6300 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR