EDTA dinatrium salt dihydrat 99+% for molekylærbiologi DNase-, RNase-, protease-free

Leverandør: Thermo Fisher Scientific
Warning

327201000 327200025 327205000
ACRO327201000EA 755 NOK
ACRO327201000 ACRO327200025 ACRO327205000
EDTA dinatrium salt dihydrat 99+% for molekylærbiologi DNase-, RNase-, protease-free
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 252 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 12,03556
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline solid
Infrared spectrum Conforms
Titration Complexometric ≥99.0 %
UV (0.1 M in water)
pH 4 to 5 (5 % at 25°C)
Dnase,Rnase,Protease activity none detected
Iron (Fe) ≤100 ppm
Heavy metals (as Pb) ≤10 ppm
Nitrilotriacetic acid ≤0.1 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR