EDTA tetrasodium salt tetrahydrate 99.5% for molecular biology DNase-, RNase-, protease-free

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: EDTA tetrasodium tetrahydrate, Tetrasodium ethylenediaminetetraacetate tetrahydrate

327211000
ACRO327211000EA 925 NOK
ACRO327211000
EDTA tetrasodium salt tetrahydrate 99.5% for molecular biology DNase-, RNase-, protease-free
EDTA tetrasodium salt tetrahydrate
Formel: C₁₀H₁₂N₂O₈Na₄·4H₂O
Smeltepunkt: > 300 °C
Tetthet: 6,9 g/cm³ (20 °C)
Flash punkt: >100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150027
CAS-nummer: 13235-36-4
EINECS: 200-573-9
Merck Indeks: 12,03557

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Titration Complexometric ≥99.4 %
Solubility (0.1 M in water) Clear colorless
UV at 280 nm A: ≤0.06
pH 10.7 to 11.7 (0.1 M in water)
Dnase,Rnase,Protease activity Not detected
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤10 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR