ACES 99+% for biochemistry

Leverandør: Thermo Fisher Scientific

Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

327671000 327670010 327672500
ACRO327671000EA 2200 NOK
ACRO327671000 ACRO327670010 ACRO327672500
ACES 99+% for biochemistry
ACES
Formel: H₂NCOCH₂NHCH₂CH₂SO₃H
Smeltepunkt: 270…295 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008030
CAS-nummer: 7365-82-4
EINECS: 230-908-4
Merck Indeks: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Heavy metals ≤5 ppm
UV at 280 nm A: ≤0.02
pH 3.6 to 4.4 (1 % at 25°C)
Titration with NaOH ≥99.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR