Calcimycin (A23187) Free_acid 98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Calcium Ionophore A23187, Calcium Ionophore III, Antibiotic A23187

328080010 328080050
ACRO328080010EA 800 NOK
ACRO328080010 ACRO328080050
Calcimycin (A23187) Free_acid 98%
Calcimycin (A23187)

CAS No.: 52665-69-7

Formel: C₂₉H₃₇N₃O₆
MW: 523.63 g/mol
Smeltepunkt: 182…193 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00151202
CAS-nummer: 52665-69-7
FN: 2811
ADR: 6.1 III

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 182 to 193 °C
TLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR