DETA NONOate (NOC-18) 95%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene

328650250 328651000
ACRO328650250EA 1125 NOK
ACRO328650250 ACRO328651000
DETA NONOate (NOC-18) 95%
DETA NONOate (NOC-18)

CAS No.: 146724-94-9

Formel: C₄H₁₃N₅O₂
Smeltepunkt: 98…106 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00278808
CAS-nummer: 146724-94-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
NMR ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR