Polysorbat 40

Leverandør: Thermo Fisher Scientific

334140010 334140250 334142500 334140025
ACRO334140010EA 1200 NOK
ACRO334140010 ACRO334140250 ACRO334142500 ACRO334140025
Polysorbat 40
Polysorbat 40
Formel: C₆₂H₁₂₂O₂₆
Kokepunkt: ? 100 °C (1013 hPa)
Smeltepunkt: ~ 0,1 °C
Tetthet: 1,05 g/cm³
Flash punkt: >200 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00165345
CAS-nummer: 9005-66-7

Bestill nå

Spesifikasjon Testresultater

Appearance Clear to hazy light yellow to dark yellow Viscous liquid or paste
Infrared spectrum Conforms
Acid value ≤3 mg KOH/g
Hydroxyl value 89 to 105 mg KOH/g
Saponification value 41 to 55 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR