Polysorbat 85

Leverandør: Thermo Fisher Scientific

334150010 334152500 334150025
ACRO334150010EA 1650 NOK
ACRO334150010 ACRO334152500 ACRO334150025
Polysorbat 85
Polysorbat 85
Tetthet: 1,02 g/cm³
Flash punkt: >200 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01779641
CAS-nummer: 9005-70-3

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to amber Viscous liquid
Infrared spectrum Conforms
Water ≤3 %
Acid value ≤2 mg KOH/g
Hydroxyl value 38 to 52 mg KOH/g
Saponification value 80 to 95 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR