4-Hydroksy-2-metoksybensaldehyd 98%

Leverandør: Thermo Fisher Scientific
Warning

335930010 335930050 335930250
ACRO335930010EA 555 NOK
ACRO335930010 ACRO335930050 ACRO335930250
4-Hydroksy-2-metoksybensaldehyd 98%
4-Hydroksy-2-metoksybensaldehyd
Formel: C₈H₈O₃
Kokepunkt: 273 °C (1013 hPa)
Smeltepunkt: 153…158 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00051964
CAS-nummer: 18278-34-7

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
Melting point 156 to 161 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR