3-Nitrophenyl isocyanate 97%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 1-Isocyanato-3-nitrobenzene, 3-Nitrophenylisocyanate

336180050 336180250
ACRO336180050EA 775 NOK
ACRO336180050 ACRO336180250
3-Nitrophenyl isocyanate 97%
3-Nitrophenyl isocyanate
Formel: C₇H₄N₂O₃
MW: 164.12 g/mol
Kokepunkt: 130…131 °C (11 mmHg)
Smeltepunkt: 50…52 °C
Flash punkt: >110 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007220
CAS-nummer: 3320-87-4
EINECS: 222-025-8
FN: 2811
ADR: 6.1 III

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 49 to 54 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR