2-brom-5-nitrobenso syre

Leverandør: Thermo Fisher Scientific
Warning

342290050
ACRO342290050EA 850 NOK
ACRO342290050
2-brom-5-nitrobenso syre
2-brom-5-nitrobenso syre
Formel: C₇H₄BrNO₄
Kokepunkt: 371 °C (1013 hPa)
Smeltepunkt: 180…181 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134558
CAS-nummer: 943-14-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light beige to light brown Powder
Infrared spectrum Conforms
Melting point 178 to 184 °C
HPLC ≥96.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR