3-Methylfuran 98%

Leverandør: Thermo Fisher Scientific
Danger

342300010
ACRO342300010EA 2950 NOK
ACRO342300010
3-Methylfuran 98%
3-Methylfuran
Formel: C₅H₆O
Kokepunkt: 65…66 °C
Tetthet: 0,92 g/cm³
Flash punkt: –22 °C
MDL nummer: MFCD00060134
CAS-nummer: 930-27-8
FN: 1992
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
GC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR