N,N-Dimethylglycine 97%

Leverandør: Thermo Fisher Scientific
Warning

345200050 345200250
ACRO345200050EA 1300 NOK
ACRO345200050 ACRO345200250
N,N-Dimethylglycine 97%
N,N-Dimethylglycine
Formel: C₄H₉NO₂
Smeltepunkt: 175…181 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004283
CAS-nummer: 1118-68-9
EINECS: 214-267-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 175 to 181 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR