2-Methylpyridin-3-ol 99%

Leverandør: Thermo Scientific
Danger

Synonymer: 3-Hydroxy-2-methylpyridine

345220050
ACRO345220050EA 705 NOK
ACRO345220050
2-Methylpyridin-3-ol 99%
2-Methylpyridin-3-ol
Formel: C₆H₇NO
MW: 109,13 g/mol
Smeltepunkt: 167…171 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082538
CAS-nummer: 1121-25-1
EINECS: 214-327-3

Bestill nå

Spesifikasjon Testresultater

Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 167 to 171 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR