Carminic acid 70-90%

Leverandør: Thermo Fisher Scientific
Danger

346630010 346630050
ACRO346630010EA 1400 NOK
ACRO346630010 ACRO346630050
Carminic acid 70-90%
Carminic acid
Formel: C₂₂H₂₀O₁₃
Smeltepunkt: 136 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167028
CAS-nummer: 1260-17-9
EINECS: 215-023-3
FN: 3147
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Red Crystalline powder
Infrared spectrum Conforms
HPLC ≥70 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR