Ferrocenemethanol 97%

Leverandør: Thermo Fisher Scientific
Warning

348030010 348030050 348035000
ACRO348030010EA 1700 NOK
ACRO348030010 ACRO348030050 ACRO348035000
Ferrocenemethanol 97%
Ferrocenemethanol
Formel: C₁₁H₁₂FeO
Smeltepunkt: 77…82 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00040878
CAS-nummer: 1273-86-5

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to golden to orange Powder or needles
Infrared spectrum Conforms
Melting point 77 to 82 °C
HPLC ≥96.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR