Toluidinblått O, pure

Leverandør: Thermo Fisher Scientific

Synonymer: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

348600050 348600250 348601000
ACRO348600050EA 710 NOK
ACRO348600050 ACRO348600250 ACRO348601000
Toluidinblått O, pure
Toluidinblått O
Formel: C₁₅H₁₆ClN₃S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011934
CAS-nummer: 92-31-9
EINECS: 202-146-2
Merck Indeks: 12,09658

Bestill nå

Spesifikasjon Testresultater

Appearance Dark green Powder
Infrared spectrum Conforms
UV spectrum Complies

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR